siRNA Details
Gene Symbol: Olfr1428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146678.2 | NP_666889.2 | 1 | 287 | ||
GenBank mRNA | BC119441.2 | - | 1 | 287 | 40042424 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146678.2 | NP_666889.2 | 1 | 287 | ||
GenBank mRNA | BC119441.2 | - | 1 | 287 | 40042424 |