siRNA Details
Gene Symbol: Olfr1427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146679.1 | NP_666890.1 | 1 | 823 | ||
GenBank mRNA | BC127962.1 | - | 1 | 827 | 40134881 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
