siRNA Details
| Gene Symbol: Olfr76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146682.1 | NP_666893.1 | 1 | 811 | ||
| GenBank mRNA | BC138072.1 | - | 1 | 837 | 8861091 | |
| BC138073.1 | - | 1 | 837 | 8861092 | ||
| Y15525.1 | - | 1 | 625 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr76| Gene Symbol: Olfr76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146682.1 | NP_666893.1 | 1 | 811 | ||
| GenBank mRNA | BC138072.1 | - | 1 | 837 | 8861091 | |
| BC138073.1 | - | 1 | 837 | 8861092 | ||
| Y15525.1 | - | 1 | 625 | |||