siRNA Details
| Gene Symbol: Olfr1431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146414.1 | NP_666526.1 | 1 | 548 | ||
| GenBank mRNA | BC133710.1 | - | 1 | 552 | 40142241 | |
| Gene Symbol: Olfr235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146686.2 | NP_666897.2 | 1 | 548 | ||
| GenBank mRNA | BC132238.1 | - | 1 | 618 | 40130515 | |
| BC132388.1 | - | 1 | 618 | 40130665 | ||

Olfr1431
Genomic Map