siRNA Details
Gene Symbol: Olfr1436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146687.2 | NP_666898.2 | 1 | 786 | ||
GenBank mRNA | BC127991.1 | - | 1 | 790 | 40135681 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146687.2 | NP_666898.2 | 1 | 786 | ||
GenBank mRNA | BC127991.1 | - | 1 | 790 | 40135681 |