siRNA Details
| Gene Symbol: Olfr1459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146689.1 | NP_666900.1 | 1 | 578 | ||
| GenBank mRNA | BC138407.1 | - | 1 | 664 | 8861427 | |
| BC138408.1 | - | 1 | 664 | 8861428 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1459| Gene Symbol: Olfr1459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146689.1 | NP_666900.1 | 1 | 578 | ||
| GenBank mRNA | BC138407.1 | - | 1 | 664 | 8861427 | |
| BC138408.1 | - | 1 | 664 | 8861428 | ||