siRNA Details
| Gene Symbol: Olfr1453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146700.1 | NP_666911.1 | 1 | 109 | ||
| Gene Symbol: Olfr1454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146692.1 | NP_666903.1 | 1 | 109 | ||

Olfr1453
Genomic Map