siRNA Details
| Gene Symbol: Olfr1444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146702.1 | NP_666913.1 | 1 | 595 | ||
| GenBank mRNA | BC119059.1 | - | 1 | 630 | 8733808 | |
| BC125619.1 | - | 1 | 630 | 40130134 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1444| Gene Symbol: Olfr1444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146702.1 | NP_666913.1 | 1 | 595 | ||
| GenBank mRNA | BC119059.1 | - | 1 | 630 | 8733808 | |
| BC125619.1 | - | 1 | 630 | 40130134 | ||