siRNA Details
| Gene Symbol: Olfr1447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146703.1 | NP_666914.1 | 1 | 451 | ||
| GenBank mRNA | BC125359.1 | - | 1 | 534 | 40129874 | |
| BC132255.1 | - | 1 | 534 | 40130532 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1447| Gene Symbol: Olfr1447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146703.1 | NP_666914.1 | 1 | 451 | ||
| GenBank mRNA | BC125359.1 | - | 1 | 534 | 40129874 | |
| BC132255.1 | - | 1 | 534 | 40130532 | ||