siRNA Details
| Gene Symbol: Olfr410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146707.1 | NP_666918.1 | 1 | 813 | ||
| Gene Symbol: Olfr401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146706.1 | NP_666917.1 | 1 | 813 | ||

Olfr410
Genomic Map