siRNA Details
| Gene Symbol: Olfr411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146709.2 | NP_666920.2 | 1 | 250 | ||
| GenBank mRNA | BC138439.1 | - | 1 | 250 | 8861459 | |
| BC138440.1 | - | 1 | 250 | 8861460 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr411| Gene Symbol: Olfr411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146709.2 | NP_666920.2 | 1 | 250 | ||
| GenBank mRNA | BC138439.1 | - | 1 | 250 | 8861459 | |
| BC138440.1 | - | 1 | 250 | 8861460 | ||