siRNA Details
| Gene Symbol: Olfr432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146716.2 | NP_666927.2 | 1 | 739 | ||
| GenBank mRNA | BC106792.1 | - | 1 | 737 | ||
| BC106793.2 | - | 1 | 739 | 40041669 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr432| Gene Symbol: Olfr432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146716.2 | NP_666927.2 | 1 | 739 | ||
| GenBank mRNA | BC106792.1 | - | 1 | 737 | ||
| BC106793.2 | - | 1 | 739 | 40041669 | ||