siRNA Details
| Gene Symbol: PRSS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002773.4 | NP_002764.1 | 3 | 446 | ||
| GenBank mRNA | AU142128.1 | - | 3 | 446 | ||
| BC001462.2 | - | 3 | 357 | 3138532 | ||
| L41351.1 | - | 3 | 410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS8| Gene Symbol: PRSS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002773.4 | NP_002764.1 | 3 | 446 | ||
| GenBank mRNA | AU142128.1 | - | 3 | 446 | ||
| BC001462.2 | - | 3 | 357 | 3138532 | ||
| L41351.1 | - | 3 | 410 | |||