siRNA Details
| Gene Symbol: Olfr1095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146730.2 | NP_666941.2 | 1 | 107 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1095| Gene Symbol: Olfr1095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146730.2 | NP_666941.2 | 1 | 107 | ||