siRNA Details
| Gene Symbol: Olfr494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146737.1 | NP_666948.1 | 1 | 660 | ||
| Gene Symbol: Olfr497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146738.1 | NP_666949.1 | 1 | 660 | ||

Olfr494
Genomic Map