siRNA Details
| Gene Symbol: Olfr1497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146741.1 | NP_666952.1 | 1 | 165 | ||
| GenBank mRNA | BC111435.1 | - | 1 | 163 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1497| Gene Symbol: Olfr1497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146741.1 | NP_666952.1 | 1 | 165 | ||
| GenBank mRNA | BC111435.1 | - | 1 | 163 | ||