siRNA Details
| Gene Symbol: Olfr935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146746.1 | NP_666957.1 | 1 | 55 | ||
| Gene Symbol: Olfr146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146747.1 | NP_666958.1 | 1 | 55 | ||
| GenBank mRNA | BC132188.1 | - | 1 | 135 | 40130465 | |
| BC132190.1 | - | 1 | 135 | 40130467 | ||

Olfr935
Genomic Map