siRNA Details
| Gene Symbol: Olfr874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146882.2 | NP_667093.2 | 1 | 197 | ||
| Gene Symbol: Olfr875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146749.2 | NP_666960.2 | 1 | 221 | ||
| GenBank mRNA | BC132237.1 | - | 1 | 221 | 40130514 | |
| BC138017.1 | - | 1 | 221 | 8861036 | ||

Olfr874
Genomic Map