siRNA Details
| Gene Symbol: Olfr648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146751.1 | NP_666962.1 | 1 | 625 | ||
| GenBank mRNA | BC106984.2 | - | 1 | 625 | 40043237 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr648| Gene Symbol: Olfr648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146751.1 | NP_666962.1 | 1 | 625 | ||
| GenBank mRNA | BC106984.2 | - | 1 | 625 | 40043237 | |