siRNA Details
| Gene Symbol: Olfr1106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146752.2 | NP_666963.2 | 1 | 139 | ||
| GenBank mRNA | BC127974.1 | - | 1 | 143 | 40135141 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1106| Gene Symbol: Olfr1106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146752.2 | NP_666963.2 | 1 | 139 | ||
| GenBank mRNA | BC127974.1 | - | 1 | 143 | 40135141 | |