siRNA Details
| Gene Symbol: Olfr608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146756.2 | NP_666967.2 | 1 | 646 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr608| Gene Symbol: Olfr608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146756.2 | NP_666967.2 | 1 | 646 | ||