siRNA Details
| Gene Symbol: Olfr583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146757.1 | NP_666968.1 | 1 | 420 | ||
| Gene Symbol: Olfr585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC145897.1 | - | 1 | 443 | 40131200 | |
More Information
Set Membership: |
|

Olfr583
Genomic Map