siRNA Details
| Gene Symbol: Olfr765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085477.1 | NP_001078946.1 | 1 | 638 | ||
| Gene Symbol: Olfr767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146318.2 | NP_666430.2 | 1 | 638 | ||
| GenBank mRNA | BC137767.1 | - | 1 | 734 | 8860786 | |

Olfr765
Genomic Map