siRNA Details
| Gene Symbol: Olfr767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146318.2 | NP_666430.2 | 1 | 881 | ||
| GenBank mRNA | BC137767.1 | - | 1 | 977 | 8860786 | |
| Gene Symbol: Olfr765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085477.1 | NP_001078946.1 | 1 | 881 | ||

Olfr767
Genomic Map