siRNA Details
| Gene Symbol: Olfr259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146770.2 | NP_666981.2 | 1 | 322 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr259| Gene Symbol: Olfr259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146770.2 | NP_666981.2 | 1 | 322 | ||