siRNA Details
| Gene Symbol: Olfr472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146774.1 | NP_666985.1 | 1 | 517 | ||
| GenBank mRNA | BC138079.1 | - | 1 | 609 | 8861098 | |
| BC145756.1 | - | 1 | 609 | 40131059 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr472| Gene Symbol: Olfr472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146774.1 | NP_666985.1 | 1 | 517 | ||
| GenBank mRNA | BC138079.1 | - | 1 | 609 | 8861098 | |
| BC145756.1 | - | 1 | 609 | 40131059 | ||