siRNA Details
| Gene Symbol: Olfr1208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146778.1 | NP_666989.1 | 1 | 127 | ||
| GenBank mRNA | BC150697.1 | - | 1 | 180 | 9087608 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1208| Gene Symbol: Olfr1208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146778.1 | NP_666989.1 | 1 | 127 | ||
| GenBank mRNA | BC150697.1 | - | 1 | 180 | 9087608 | |