siRNA Details
| Gene Symbol: Olfr915 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146785.2 | NP_666996.2 | 1 | 256 | ||
| GenBank mRNA | BC111098.1 | - | 1 | 251 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr915| Gene Symbol: Olfr915 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146785.2 | NP_666996.2 | 1 | 256 | ||
| GenBank mRNA | BC111098.1 | - | 1 | 251 | ||