siRNA Details
| Gene Symbol: Olfr1230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146789.1 | NP_667000.1 | 1 | 49 | ||
| GenBank mRNA | BC108937.1 | - | 1 | 49 | 40040283 | |
| BC108938.1 | - | 1 | 49 | 40040284 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1230| Gene Symbol: Olfr1230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146789.1 | NP_667000.1 | 1 | 49 | ||
| GenBank mRNA | BC108937.1 | - | 1 | 49 | 40040283 | |
| BC108938.1 | - | 1 | 49 | 40040284 | ||