siRNA Details
| Gene Symbol: Olfr1263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146794.1 | NP_667005.1 | 1 | 433 | ||
| GenBank mRNA | BC119193.1 | - | 1 | 474 | 8733942 | |
| BC119197.1 | - | 1 | 474 | 8733946 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1263| Gene Symbol: Olfr1263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146794.1 | NP_667005.1 | 1 | 433 | ||
| GenBank mRNA | BC119193.1 | - | 1 | 474 | 8733942 | |
| BC119197.1 | - | 1 | 474 | 8733946 | ||