siRNA Details
| Gene Symbol: Olfr1502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146797.1 | NP_667008.1 | 1 | 85 | ||
| GenBank mRNA | BC106810.1 | - | 1 | 85 | 40042985 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1502| Gene Symbol: Olfr1502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146797.1 | NP_667008.1 | 1 | 85 | ||
| GenBank mRNA | BC106810.1 | - | 1 | 85 | 40042985 | |