siRNA Details
| Gene Symbol: Olfr878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146798.2 | NP_667009.1 | 1 | 628 | ||
| GenBank mRNA | BC119219.1 | - | 1 | 626 | 8733968 | |
| BC119221.1 | - | 1 | 628 | 8733970 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr878| Gene Symbol: Olfr878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146798.2 | NP_667009.1 | 1 | 628 | ||
| GenBank mRNA | BC119219.1 | - | 1 | 626 | 8733968 | |
| BC119221.1 | - | 1 | 628 | 8733970 | ||