siRNA Details
Gene Symbol: Olfr904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146801.2 | NP_667012.2 | 1 | 110 | ||
GenBank mRNA | BC114635.1 | - | 1 | 109 | ||
BC146614.1 | - | 1 | 110 | 40040862 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146801.2 | NP_667012.2 | 1 | 110 | ||
GenBank mRNA | BC114635.1 | - | 1 | 109 | ||
BC146614.1 | - | 1 | 110 | 40040862 |