siRNA Details
| Gene Symbol: Olfr136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146807.1 | NP_667018.1 | 1 | 333 | ||
| Gene Symbol: Olfr138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130868.1 | NP_570938.1 | 1 | 333 | ||
| GenBank mRNA | BC104280.1 | - | 1 | 333 | 40041704 | |
| BC104281.1 | - | 1 | 333 | 40041709 | ||
| BC127129.1 | - | 1 | 333 | 40041712 | ||
| BC127966.1 | - | 1 | 337 | 40134997 | ||

Olfr136
Genomic Map