siRNA Details
| Gene Symbol: Olfr912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146810.2 | NP_667021.2 | 2 | 356 | ||
| GenBank mRNA | CB174340.1 | - | 1 | 30 | ||
| CB174492.1 | - | 2 | 356 | |||
| Gene Symbol: Olfr910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146811.2 | NP_667022.1 | 2 | 333 | ||

Olfr912
Genomic Map