siRNA Details
| Gene Symbol: Olfr1156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146817.2 | NP_667028.2 | 1 | 327 | ||
| GenBank mRNA | BC116852.1 | - | 1 | 327 | 40126171 | |
| BC116854.1 | - | 1 | 327 | 40126173 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1156| Gene Symbol: Olfr1156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146817.2 | NP_667028.2 | 1 | 327 | ||
| GenBank mRNA | BC116852.1 | - | 1 | 327 | 40126171 | |
| BC116854.1 | - | 1 | 327 | 40126173 | ||