siRNA Details
Gene Symbol: Olfr655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146820.2 | NP_667031.2 | 1 | 855 | ||
GenBank mRNA | BC114416.1 | - | 1 | 854 | 40040785 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146820.2 | NP_667031.2 | 1 | 855 | ||
GenBank mRNA | BC114416.1 | - | 1 | 854 | 40040785 |