siRNA Details
Gene Symbol: Olfr629 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146821.2 | NP_667032.2 | 1 | 727 | ||
GenBank mRNA | BC120634.1 | - | 1 | 727 | 40129557 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr629 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146821.2 | NP_667032.2 | 1 | 727 | ||
GenBank mRNA | BC120634.1 | - | 1 | 727 | 40129557 |