siRNA Details
| Gene Symbol: Olfr969 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146826.1 | NP_667037.1 | 1 | 46 | ||
| Gene Symbol: Olfr970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146611.1 | NP_666822.1 | 1 | 46 | ||

Olfr969
Genomic Map