siRNA Details
| Gene Symbol: PSG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021016.3 | NP_066296.2 | 2 | 386 | ||
| XM_011527126.1 | XP_011525428.1 | 2 | 386 | |||
| XM_011527127.1 | XP_011525429.1 | 2 | 386 | |||
| GenBank mRNA | BC005924.1 | - | 2 | 351 | 4043628 | |
| DA833589.1 | - | 2 | 386 | |||
| M23575.1 | - | 2 | 485 | |||
| M30628.1 | - | 3 | 235 | |||
| M34420.1 | - | 2 | 335 | |||
| M37399.1 | - | 2 | 340 | |||
| M93061.1 | - | 2 | 312 | |||
| S59493.1 | - | 2 | 312 | |||
| S59494.1 | - | 3 | 241 | |||

PSG3
Genomic Map