siRNA Details
| Gene Symbol: Olfr975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146828.1 | NP_667039.1 | 1 | 814 | ||
| GenBank mRNA | BC100677.1 | - | 1 | 813 | ||
| BC101947.1 | - | 1 | 813 | |||
| BC141889.1 | - | 1 | 812 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr975| Gene Symbol: Olfr975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146828.1 | NP_667039.1 | 1 | 814 | ||
| GenBank mRNA | BC100677.1 | - | 1 | 813 | ||
| BC101947.1 | - | 1 | 813 | |||
| BC141889.1 | - | 1 | 812 | |||