siRNA Details
| Gene Symbol: Olfr133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146831.1 | NP_667042.1 | 1 | 794 | ||
| GenBank mRNA | BC132057.1 | - | 1 | 883 | 40130334 | |
| BC132059.1 | - | 1 | 883 | 40130336 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr133| Gene Symbol: Olfr133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146831.1 | NP_667042.1 | 1 | 794 | ||
| GenBank mRNA | BC132057.1 | - | 1 | 883 | 40130334 | |
| BC132059.1 | - | 1 | 883 | 40130336 | ||