siRNA Details
| Gene Symbol: Olfr101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146834.1 | NP_667045.1 | 1 | 49 | ||
| GenBank mRNA | BC138322.1 | - | 1 | 113 | 8861342 | |
| BC138323.1 | - | 1 | 113 | 8861343 | ||
| Gene Symbol: Olfr102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011721.2 | NP_001011721.2 | 1 | 104 | ||
| GenBank mRNA | BC139114.1 | - | 1 | 104 | 8862136 | |
| BC139115.1 | - | 1 | 104 | 8862137 | ||

Olfr101
Genomic Map