siRNA Details
Gene Symbol: Olfr1098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146845.2 | NP_667056.2 | 1 | 608 | ||
GenBank mRNA | BC113138.1 | - | 1 | 586 |
Gene Symbol: Olfr1097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146843.2 | NP_667054.2 | 2 | 883 | ||
GenBank mRNA | BC104098.1 | - | 1 | 608 | 40043068 | |
BC104109.1 | - | 1 | 606 | |||
BC106801.1 | - | 1 | 608 | 40042504 |
