siRNA Details
| Gene Symbol: Olfr1107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146844.2 | NP_667055.2 | 1 | 498 | ||
| GenBank mRNA | BC120852.1 | - | 1 | 498 | 40129775 | |
| BC137830.1 | - | 1 | 498 | 8860849 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1107| Gene Symbol: Olfr1107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146844.2 | NP_667055.2 | 1 | 498 | ||
| GenBank mRNA | BC120852.1 | - | 1 | 498 | 40129775 | |
| BC137830.1 | - | 1 | 498 | 8860849 | ||