siRNA Details
| Gene Symbol: Olfr1098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146845.2 | NP_667056.2 | 1 | 218 | ||
| GenBank mRNA | BC113138.1 | - | 1 | 196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1098| Gene Symbol: Olfr1098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146845.2 | NP_667056.2 | 1 | 218 | ||
| GenBank mRNA | BC113138.1 | - | 1 | 196 | ||