siRNA Details
| Gene Symbol: Olfr1339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146852.2 | NP_667063.1 | 1 | 351 | ||
| GenBank mRNA | BC104296.1 | - | 1 | 350 | ||
| BC104297.1 | - | 1 | 350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1339| Gene Symbol: Olfr1339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146852.2 | NP_667063.1 | 1 | 351 | ||
| GenBank mRNA | BC104296.1 | - | 1 | 350 | ||
| BC104297.1 | - | 1 | 350 | |||