siRNA Details
| Gene Symbol: PSG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130014.1 | NP_001123486.1 | 2 | 307 | ||
| NM_002781.3 | NP_002772.3 | 2 | 307 | |||
| XM_011527132.2 | XP_011525434.1 | 2 | 305 | |||
| XM_017027003.1 | XP_016882492.1 | 2 | 305 | |||
| GenBank mRNA | BC012607.1 | - | 2 | 274 | 4043912 | |
| BG622527.1 | - | 2 | 275 | |||
| DA835167.1 | - | 2 | 307 | |||
| M25384.1 | - | 2 | 275 | |||
| M73713.1 | - | 2 | 261 | |||

PSG5
Genomic Map