siRNA Details
| Gene Symbol: Olfr1341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146853.2 | NP_667064.2 | 1 | 616 | ||
| GenBank mRNA | BC130239.1 | - | 1 | 620 | ||
| BC152333.1 | - | 1 | 620 | |||
| BC152353.1 | - | 1 | 620 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1341| Gene Symbol: Olfr1341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146853.2 | NP_667064.2 | 1 | 616 | ||
| GenBank mRNA | BC130239.1 | - | 1 | 620 | ||
| BC152333.1 | - | 1 | 620 | |||
| BC152353.1 | - | 1 | 620 | |||