siRNA Details
| Gene Symbol: PSG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031850.3 | NP_001027020.1 | 3 | 666 | ||
| NM_002782.4 | NP_002773.1 | 3 | 666 | |||
| GenBank mRNA | BC020652.1 | - | 3 | 633 | 4770562 | |
| M31125.1 | - | 3 | 579 | |||
| M33666.1 | - | 3 | 586 | |||
| X17098.1 | - | 3 | 587 | |||

PSG6
Genomic Map